Alfa Aesar™ 4-Hydroxy-3-methylpyridine, 97%

Manufacturer: Alfa Aesar™ H3226506

Quantity 5g

 View more versions of this product

Catalog No. AAH3226506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22280-02-0
    Molecular Formula C6H7NO
    Formula Weight 109.13
    MDL Number MFCD04114239
    Synonym 4-Hydroxy-2-picoline; 3-Methyl-4-pyridinol
    Chemical Name or Material 4-Hydroxy-3-methylpyridine

    Specifications

    Formula Weight 109.13
    Molecular Formula C6H7NO
    MDL Number MFCD04114239
    Synonym 4-Hydroxy-2-picoline; 3-Methyl-4-pyridinol
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point ≈168°C
    Quantity 5g
    CAS 22280-02-0
    Health Hazard 1 Warning
    Chemical Name or Material 4-Hydroxy-3-methylpyridine
    Assay 97%

    Description

    Applications
    4-Hydroxy-3-methylpyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Soluble in water (14 g/L) (25°C).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit