Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrobenzenearsonic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
121-19-7
C6H6AsNO6
263.037
MFCD00007112
XMVJITFPVVRMHC-UHFFFAOYSA-N
roxarsone,3-nitro,ristat,4-hydroxy-3-nitrobenzenearsonic acid,aklomix-3,roxarson,ren-o-sal,3-nitro-10,3n4hpa,nitrophenolarsonic acid
5104
CHEBI:35817
(4-hydroxy-3-nitrophenyl)arsonic acid
C1=CC(=C(C=C1[As](=O)(O)O)[N+](=O)[O-])O
121-19-7
C6H6AsNO6
263.037
MFCD00007112
XMVJITFPVVRMHC-UHFFFAOYSA-N
roxarsone,3-nitro,ristat,4-hydroxy-3-nitrobenzenearsonic acid,aklomix-3,roxarson,ren-o-sal,3-nitro-10,3n4hpa,nitrophenolarsonic acid
5104
CHEBI:35817
(4-hydroxy-3-nitrophenyl)arsonic acid
C1=CC(=C(C=C1[As](=O)(O)O)[N+](=O)[O-])O
Water 1% max.
Color Beige-Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H3(NO2)AsO3H2
Beilstein 16, I, 456
Merck Index 15, 8406
Formula Weight 263.03
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material 4-Hydroxy-3-nitrobenzenearsonic acid, 98%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-453-7

RTECSNumber : CY5250000

TSCA : TSCA