Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrobenzonitrile 97%, ACROS Organics™

Click to view available options
Quantity:
10g
3272-08-0
C7H4N2O3
164.12
MFCD00070775
INBLGVOPOSGVTA-UHFFFAOYSA-N
4-hydroxy-3-nitrobenzonitrile,2-nitro-4-cyanophenol,benzonitrile, 4-hydroxy-3-nitro,3-nitro-4-hydroxybenzonitrile,4-cyano-2-nitrophenol,4-hydroxy-3-nitro-benzonitrile,4-hydroxy-3-nitrobenzenecarbonitrile,pubchem19690,acmc-1cq5c,ksc494q7b
76758
C1=CC(=C(C=C1C#N)[N+](=O)[O-])O
3272-08-0
C7H4N2O3
164.12
MFCD00070775
INBLGVOPOSGVTA-UHFFFAOYSA-N
4-hydroxy-3-nitrobenzonitrile,2-nitro-4-cyanophenol,benzonitrile, 4-hydroxy-3-nitro,3-nitro-4-hydroxybenzonitrile,4-cyano-2-nitrophenol,4-hydroxy-3-nitro-benzonitrile,4-hydroxy-3-nitrobenzenecarbonitrile,pubchem19690,acmc-1cq5c,ksc494q7b
76758
C1=CC(=C(C=C1C#N)[N+](=O)[O-])O
Quantity 10g
Linear Formula NO2C6H3(OH)CN
Formula Weight 164.12
Chemical Name or Material 4-Hydroxy-3-nitrobenzonitrile, 97%