Alfa Aesar™ 4-Hydroxy-3-nitrocoumarin, 98%

Manufacturer: Alfa Aesar™ A1222122

Quantity 100g

 View more versions of this product

Catalog No. AAA1222122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20261-31-8
    Molecular Formula C9H5NO5
    Formula Weight 207.14
    MDL Number MFCD00051671
    Beilstein 1247343
    Synonym 4-Hydroxy-3-nitro-2H-chromen-2-one
    Chemical Name or Material 4-Hydroxy-3-nitrocoumarin

    Specifications

    Formula Weight 207.14
    MDL Number MFCD00051671
    CAS 20261-31-8
    Beilstein 1247343
    Synonym 4-Hydroxy-3-nitro-2H-chromen-2-one
    Melting Point ≈171°C (dec.)
    Quantity 100g
    Molecular Formula C9H5NO5
    Chemical Name or Material 4-Hydroxy-3-nitrocoumarin
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit