Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrocoumarin, 98%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
20261-31-8
C9H5NO5
207.141
MFCD00051671
NZQAQAUWFHMVEM-UHFFFAOYSA-N
4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
54682640
4-hydroxy-3-nitrochromen-2-one
C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O
20261-31-8
C9H5NO5
207.141
MFCD00051671
NZQAQAUWFHMVEM-UHFFFAOYSA-N
4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
54682640
4-hydroxy-3-nitrochromen-2-one
C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point ∼171°C (decomposition)
Beilstein 1247343
Quantity 5g
Formula Weight 207.14
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-nitrocoumarin

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only