Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
10463-20-4
C8H7NO5
197.15
QBHBHOSRLDPIHG-UHFFFAOYSA-N
4-hydroxy-3-nitrophenylacetic acid,2-4-hydroxy-3-nitrophenyl acetic acid,4-hydroxy-3-nitrophenyl acetic acid,3-nitro-4-hydroxyphenylacetic acid,3-nitro-4-hydroxy phenylacetic acid,benzeneacetic acid, 4-hydroxy-3-nitro,no2hpa,hydroxynitrophenylaceticacid,acmc-2098cj,epitope id:158629
447364
CHEBI:546274
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O
10463-20-4
C8H7NO5
197.15
QBHBHOSRLDPIHG-UHFFFAOYSA-N
4-hydroxy-3-nitrophenylacetic acid,2-4-hydroxy-3-nitrophenyl acetic acid,4-hydroxy-3-nitrophenyl acetic acid,3-nitro-4-hydroxyphenylacetic acid,3-nitro-4-hydroxy phenylacetic acid,benzeneacetic acid, 4-hydroxy-3-nitro,no2hpa,hydroxynitrophenylaceticacid,acmc-2098cj,epitope id:158629
447364
CHEBI:546274
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O
Melting Point 144.0°C to 147.0°C
Color Colorless to Yellow
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula HOC6H3(NO2)CH2CO2H
Beilstein 10,IV,562
Formula Weight 197.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic acid, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 233-948-0