Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-(trifluoromethyl)benzoic acid, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
220239-68-9
C8H5F3O3
206.12
DPVRVZQEDJVWLS-UHFFFAOYSA-N
4-hydroxy-3-trifluoromethyl benzoic acid,benzoic acid, 4-hydroxy-3-trifluoromethyl,4-hydroxy-3-trifluoromethyl benzoicacid,acmc-1cdpd,ksc495o6p,4-hydroxy-3-trifluoromethyl-benzoic acid,4-oxidanyl-3-trifluoromethyl benzoic acid
2775112
4-hydroxy-3-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(=O)O)C(F)(F)F)O
220239-68-9
C8H5F3O3
206.12
DPVRVZQEDJVWLS-UHFFFAOYSA-N
4-hydroxy-3-trifluoromethyl benzoic acid,benzoic acid, 4-hydroxy-3-trifluoromethyl,4-hydroxy-3-trifluoromethyl benzoicacid,acmc-1cdpd,ksc495o6p,4-hydroxy-3-trifluoromethyl-benzoic acid,4-oxidanyl-3-trifluoromethyl benzoic acid
2775112
4-hydroxy-3-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(=O)O)C(F)(F)F)O
Melting Point 144.0°C to 147.0°C
Packaging Glass bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Linear Formula CF3C6H3CO2H(OH)
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning