Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxy-4-biphenylcarbonitrile, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
19812-93-2
C13H9NO
195.221
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl,4'-hydroxy-4-biphenylcarbonitrile,4-4-hydroxyphenyl benzonitrile,4'-hydroxybiphenyl-4-carbonitrile,4'-cyano-4-hydroxybiphenyl,4'-hydroxy-1,1'-biphenyl-4-carbonitrile,4'-cyano-4-biphenylol,4-cyano-4'-hydroxy-biphenyl,4'-hydroxy-4-cyanobiphenyl,4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
19812-93-2
C13H9NO
195.221
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl,4'-hydroxy-4-biphenylcarbonitrile,4-4-hydroxyphenyl benzonitrile,4'-hydroxybiphenyl-4-carbonitrile,4'-cyano-4-hydroxybiphenyl,4'-hydroxy-1,1'-biphenyl-4-carbonitrile,4'-cyano-4-biphenylol,4-cyano-4'-hydroxy-biphenyl,4'-hydroxy-4-cyanobiphenyl,4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
Melting Point 192°C to 199°C
Quantity 5g
Color Yellow to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula NC(C6H4)2OH
Formula Weight 195.22
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material 4';-Hydroxy-4-biphenylcarbonitrile, 95%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA