Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxy-4'-iodobiphenyl, 98+%

Click to view available options
Quantity:
5g
25g
100g
29558-78-9
C12H9IO
296.107
MFCD02258843
WJXIAMCDWSUSEI-UHFFFAOYSA-N
4-hydroxy-4'-iodobiphenyl,4'-iodo-1,1'-biphenyl-4-ol,4-4-iodophenyl phenol,4-4'-iodophenyl phenol,4'-iodobiphenyl-4-ol,acmc-20amgy,pubchem9061,4-4?iodophenyl phenol,4-4'-iodophenyl-phenol
2064030
4-(4-iodophenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)I)O
29558-78-9
C12H9IO
296.107
MFCD02258843
WJXIAMCDWSUSEI-UHFFFAOYSA-N
4-hydroxy-4'-iodobiphenyl,4'-iodo-1,1'-biphenyl-4-ol,4-4-iodophenyl phenol,4-4'-iodophenyl phenol,4'-iodobiphenyl-4-ol,acmc-20amgy,pubchem9061,4-4?iodophenyl phenol,4-4'-iodophenyl-phenol
2064030
4-(4-iodophenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)I)O
Quantity 5g
Melting Point 117°C to 119°C
Formula Weight 296.11
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-4'-iodobiphenyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning