Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-hydroxy-6-mercaptopyrazolo[3,4-d]pyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
24521-76-4
C5H4N4OS
168.18
MFCD00022782
SXRSXYWROQWSGJ-UHFFFAOYSA-N
4-hydroxy-6-mercaptopyrazolo 3,4-d pyrimidine,6-mercapto-1h-pyrazolo 3,4-d pyrimidin-4-ol,4-hydroxy-6-mercapto-1h-pyrazolo 3,4-d pyrimidine,6-mercapto-4-hydroxy-1h-pyrazolo 3,4-d pyrimidine,6-sulfanyl-1h,4h,5h-pyrazolo 3,4-d pyrimidin-4-one,6-mercapto-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one,1,5,6,7-tetrahydro-6-thioxo-4h-pyrazolo 3,4-d pyrimidin-4-one,4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5,6,7-tetrahydro-6-thioxo,4h-pyrazolo 3,4-d pyrimidin-4-one,1,5,6,7-tetrahydro-6-thioxo
3034349
6-sulfanylidene-1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
C1=C2C(=NC(=S)NC2=O)NN1
24521-76-4
C5H4N4OS
168.18
MFCD00022782
SXRSXYWROQWSGJ-UHFFFAOYSA-N
4-hydroxy-6-mercaptopyrazolo 3,4-d pyrimidine,6-mercapto-1h-pyrazolo 3,4-d pyrimidin-4-ol,4-hydroxy-6-mercapto-1h-pyrazolo 3,4-d pyrimidine,6-mercapto-4-hydroxy-1h-pyrazolo 3,4-d pyrimidine,6-sulfanyl-1h,4h,5h-pyrazolo 3,4-d pyrimidin-4-one,6-mercapto-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one,1,5,6,7-tetrahydro-6-thioxo-4h-pyrazolo 3,4-d pyrimidin-4-one,4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5,6,7-tetrahydro-6-thioxo,4h-pyrazolo 3,4-d pyrimidin-4-one,1,5,6,7-tetrahydro-6-thioxo
3034349
6-sulfanylidene-1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
C1=C2C(=NC(=S)NC2=O)NN1
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Melting Point >310.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Solubility Information Solubility in water: insoluble
Formula Weight 168.18
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-6-mercaptopyrazolo[3,4-d]pyrimidine
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 246-296-7