Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid, 85%, Tech., ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
574-92-5
C11H6F3NO3
257.17
MFCD00006771
BIRIVPOTERXIOW-UHFFFAOYSA-N
4-hydroxy-7-trifluoromethyl quinoline-3-carboxylic acid, 4-hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid, 4-oxo-7-trifluoromethyl-1,4-dihydroquinoline-3-carboxylic acid, 4-hydroxy-7-trifluoromethylquinoline-3-carboxylic acid, 3-quinolinecarboxylic acid, 4-hydroxy-7-trifluoromethyl, 4-oxo-7-trifluoromethyl-1h-quinoline-3-carboxylic acid, 4-22-00-02331 beilstein handbook reference, 1,4-dihydro-4-oxo-7-trifluoromethyl quinoline-3-carboxylic acid, 3-carboxy-4-hydroxy-7-trifluoromethyl quinoline
68459
4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylic acid
OC(=O)C1=CNC2=CC(=CC=C2C1=O)C(F)(F)F
574-92-5
C11H6F3NO3
257.17
MFCD00006771
BIRIVPOTERXIOW-UHFFFAOYSA-N
4-hydroxy-7-trifluoromethyl quinoline-3-carboxylic acid, 4-hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid, 4-oxo-7-trifluoromethyl-1,4-dihydroquinoline-3-carboxylic acid, 4-hydroxy-7-trifluoromethylquinoline-3-carboxylic acid, 3-quinolinecarboxylic acid, 4-hydroxy-7-trifluoromethyl, 4-oxo-7-trifluoromethyl-1h-quinoline-3-carboxylic acid, 4-22-00-02331 beilstein handbook reference, 1,4-dihydro-4-oxo-7-trifluoromethyl quinoline-3-carboxylic acid, 3-carboxy-4-hydroxy-7-trifluoromethyl quinoline
68459
4-oxo-7-(trifluoromethyl)-1,4-dihydroquinoline-3-carboxylic acid
OC(=O)C1=CNC2=CC(=CC=C2C1=O)C(F)(F)F
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
Color Beige
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 85%
Formula Weight 257.16
Physical Form Crystalline Powder
Percent Purity ≥80%
Grade Technical
Chemical Name or Material 4-Hydroxy-7-trifluoromethyl-3-quinolinecarboxylic acid, tech., 85%
Show More Show Less