Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-N-methylpiperidine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
106-52-5
C6H13NO
115.17
MFCD00006500
BAUWRHPMUVYFOD-UHFFFAOYSA-N
4-hydroxy-1-methylpiperidine,n-methyl-4-piperidinol,1-methyl-4-piperidinol,4-hydroxy-n-methylpiperidine,n-methyl-4-hydroxypiperidine,4-piperidinol, 1-methyl,1-methyl-4-hydroxypiperidine,n-methylpiperidol,n-methyl-4-hydroxy piperidine,4-hydroxy-1-methyl-piperidine
66048
1-methylpiperidin-4-ol
CN1CCC(CC1)O
106-52-5
C6H13NO
115.17
MFCD00006500
BAUWRHPMUVYFOD-UHFFFAOYSA-N
4-hydroxy-1-methylpiperidine,n-methyl-4-piperidinol,1-methyl-4-piperidinol,4-hydroxy-n-methylpiperidine,n-methyl-4-hydroxypiperidine,4-piperidinol, 1-methyl,1-methyl-4-hydroxypiperidine,n-methylpiperidol,n-methyl-4-hydroxy piperidine,4-hydroxy-1-methyl-piperidine
66048
1-methylpiperidin-4-ol
CN1CCC(CC1)O
Melting Point 29°C to 31°C
Boiling Point 200°C
Flash Point 87°C
Refractive Index 1.4760 to 1.479
Quantity 5g
Color Brown to Colorless
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, I, 188
Solubility Information Solubility in water: miscible with water
Formula Weight 115.17
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material 4-Hydroxy-N-methylpiperidine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 203-406-8

TSCA : TSCA