Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxyacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
99-93-4
C8H8O2
136.15
MFCD00002359
TXFPEBPIARQUIG-UHFFFAOYSA-N
4'-hydroxyacetophenone,4-hydroxyacetophenone,1-4-hydroxyphenyl ethanone,4-acetylphenol,p-hydroxyacetophenone,piceol,ethanone, 1-4-hydroxyphenyl,p-acetylphenol,para-hydroxyacetophenone,p-oxyacetophenone
7469
CHEBI:28032
1-(4-hydroxyphenyl)ethanone
CC(=O)C1=CC=C(C=C1)O
99-93-4
C8H8O2
136.15
MFCD00002359
TXFPEBPIARQUIG-UHFFFAOYSA-N
4'-hydroxyacetophenone,4-hydroxyacetophenone,1-4-hydroxyphenyl ethanone,4-acetylphenol,p-hydroxyacetophenone,piceol,ethanone, 1-4-hydroxyphenyl,p-acetylphenol,para-hydroxyacetophenone,p-oxyacetophenone
7469
CHEBI:28032
1-(4-hydroxyphenyl)ethanone
CC(=O)C1=CC=C(C=C1)O
Boiling Point 147°C to 148°C (3.0mmHg)
Flash Point 166°C
Melting Point 107°C to 111°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Beige to White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H4COCH3
Beilstein 08, 87
Solubility Information Solubility in water: 10g/L (22°C)
Formula Weight 136.15
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4'-Hydroxyacetophenone, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-802-8

RTECSNumber : AM8750000

TSCA : TSCA