Alfa Aesar™ 4'-Hydroxyacetophenone, 99%

Manufacturer: Alfa Aesar™ A1197422

Quantity 100g

 View more versions of this product

Catalog No. AAA1197422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-93-4
    Molecular Formula C8H8O2
    Formula Weight 136.15
    MDL Number MFCD00002359
    Beilstein 774355
    Chemical Name or Material 4'-Hydroxyacetophenone

    Specifications

    Formula Weight 136.15
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C8H8O2
    MDL Number MFCD00002359
    CAS 99-93-4
    Health Hazard 1 Danger
    RTECS Number AM8750000
    Boiling Point 286°C
    Melting Point 106° to 108°C
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 166°C (330°F)
    Quantity 100g
    Beilstein 774355
    Chemical Name or Material 4'-Hydroxyacetophenone
    Assay 99%

    Description

    Applications
    4 ft.-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.

    Solubility
    Soluble in 95% Ethanol. Slightly soluble in water.

    Notes
    Store at room temperature. Keep away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit