Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7030276
Click to view available options
Quantity:
1 g
5 g
269409-70-3
C12H17BO3
220.075
MFCD02093756
BICZJRAGTCRORZ-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
2734624
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
269409-70-3
C12H17BO3
220.075
MFCD02093756
BICZJRAGTCRORZ-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
2734624
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114°C to 117°C
Quantity 1 g
Formula Weight 220.07
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit