Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzenesulfonamide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
1576-43-8
C6H7NO3S
1730.19
MFCD00059190
DIRCLGLKRZLKHG-UHFFFAOYSA-N
p-hydroxybenzenesulfonamide,p-phenolsulfonamide,benzenesulfonamide, 4-hydroxy,4-sulfamoylphenol,benzenesulfonamide, p-hydroxy,4-hydroxyphenylsulfonamide,unii-1iit7r7vpj,p-hydroxybenzenesulphonamide,4-hydroxybenzenesulphonamide,4-hydroxybenzene-1-sulfonamide
74093
4-hydroxybenzenesulfonamide
C1=CC(=CC=C1O)S(=O)(=O)N
1576-43-8
C6H7NO3S
1730.19
MFCD00059190
DIRCLGLKRZLKHG-UHFFFAOYSA-N
p-hydroxybenzenesulfonamide,p-phenolsulfonamide,benzenesulfonamide, 4-hydroxy,4-sulfamoylphenol,benzenesulfonamide, p-hydroxy,4-hydroxyphenylsulfonamide,unii-1iit7r7vpj,p-hydroxybenzenesulphonamide,4-hydroxybenzenesulphonamide,4-hydroxybenzene-1-sulfonamide
74093
4-hydroxybenzenesulfonamide
C1=CC(=CC=C1O)S(=O)(=O)N
Quantity 50g
Melting Point 165.4°C to 168.4°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Linear Formula HOC6H4SO2NH2
Formula Weight 1730.19
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzenesulfonamide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-408-9