Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzophenone +99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
1137-42-4
C13H10O2
198.22
MFCD00002355
NPFYZDNDJHZQKY-UHFFFAOYSA-N
4-hydroxybenzophenone,4-benzoylphenol,4-hydroxyphenyl phenyl methanone,p-benzoylphenol,p-hydroxybenzophenone,methanone, 4-hydroxyphenyl phenyl,4-hydroxyphenyl phenylmethanone,benzophenone, 4-hydroxy,unii-04r2lws0ms,4'-hydroxybenzophenone
14347
CHEBI:34421
(4-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)O
1137-42-4
C13H10O2
198.22
MFCD00002355
NPFYZDNDJHZQKY-UHFFFAOYSA-N
4-hydroxybenzophenone,4-benzoylphenol,4-hydroxyphenyl phenyl methanone,p-benzoylphenol,p-hydroxybenzophenone,methanone, 4-hydroxyphenyl phenyl,4-hydroxyphenyl phenylmethanone,benzophenone, 4-hydroxy,unii-04r2lws0ms,4'-hydroxybenzophenone
14347
CHEBI:34421
(4-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)O
Color Undesignated
Packaging Glass bottle
Melting Point 132.0°C to 135.0°C
Infrared Spectrum Authentic
Quantity 1kg
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula HOC6H4COC6H5
Beilstein 08,II,184
Solubility Information Solubility in water: insoluble
Formula Weight 198.22
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxybenzophenone, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-507-1

RTECSNumber : PC4959775

TSCA : TSCA