Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Hydroxybiphenyl-4-carbonitrile, 99%

Click to view available options
Quantity:
1g
5g
19812-93-2
C13H9NO
195.221
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl,4'-hydroxy-4-biphenylcarbonitrile,4-4-hydroxyphenyl benzonitrile,4'-hydroxybiphenyl-4-carbonitrile,4'-cyano-4-hydroxybiphenyl,4'-hydroxy-1,1'-biphenyl-4-carbonitrile,4'-cyano-4-biphenylol,4-cyano-4'-hydroxy-biphenyl,4'-hydroxy-4-cyanobiphenyl,4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
19812-93-2
C13H9NO
195.221
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl,4'-hydroxy-4-biphenylcarbonitrile,4-4-hydroxyphenyl benzonitrile,4'-hydroxybiphenyl-4-carbonitrile,4'-cyano-4-hydroxybiphenyl,4'-hydroxy-1,1'-biphenyl-4-carbonitrile,4'-cyano-4-biphenylol,4-cyano-4'-hydroxy-biphenyl,4'-hydroxy-4-cyanobiphenyl,4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
Quantity 1g
Melting Point 196°C to 199°C
UN Number UN3439
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 4'-Hydroxybiphenyl-4-carbonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.