Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Hydroxyflavanone, 98%

Click to view available options
Quantity:
1g
5g
25g
6515-37-3
C15H12O3
240.258
MFCD00017705
ZLHVIYHWWQYJID-UHFFFAOYSA-N
4'-hydroxyflavanone,2-4-hydroxyphenyl chroman-4-one,4'-hydroxy flavanone,2,3-dihydro-2-4-hydroxyphenyl-4h-1-benzopyran-4-one,2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one,chembl73933,4h-1-benzopyran-4-one, 2,3-dihydro-2-4-hydroxyphenyl,flavanone, 4'-hydroxy,4-hydroxyflavanone,4'-hydroxyflavaone
165506
CHEBI:34361
2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C1C(OC2=CC=CC=C2C1=O)C3=CC=C(C=C3)O
6515-37-3
C15H12O3
240.258
MFCD00017705
ZLHVIYHWWQYJID-UHFFFAOYSA-N
4'-hydroxyflavanone,2-4-hydroxyphenyl chroman-4-one,4'-hydroxy flavanone,2,3-dihydro-2-4-hydroxyphenyl-4h-1-benzopyran-4-one,2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one,chembl73933,4h-1-benzopyran-4-one, 2,3-dihydro-2-4-hydroxyphenyl,flavanone, 4'-hydroxy,4-hydroxyflavanone,4'-hydroxyflavaone
165506
CHEBI:34361
2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C1C(OC2=CC=CC=C2C1=O)C3=CC=C(C=C3)O
Quantity 25g
Melting Point ~182°C
Formula Weight 240.25
Percent Purity 98%
Chemical Name or Material 4'-Hydroxyflavanone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning