Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxyindole-6-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
40990-52-1
C9H7NO3
177.159
MFCD04973988
TXZOAKBAZXCPGT-UHFFFAOYSA-N
4-hydroxy-6-indolecarboxylic acid,4-hydroxyindole-6-carboxylic acid,1h-indole-6-carboxylic acid, 4-hydroxy
22742930
4-hydroxy-1H-indole-6-carboxylic acid
C1=CNC2=C1C(=CC(=C2)C(=O)O)O
40990-52-1
C9H7NO3
177.159
MFCD04973988
TXZOAKBAZXCPGT-UHFFFAOYSA-N
4-hydroxy-6-indolecarboxylic acid,4-hydroxyindole-6-carboxylic acid,1h-indole-6-carboxylic acid, 4-hydroxy
22742930
4-hydroxy-1H-indole-6-carboxylic acid
C1=CNC2=C1C(=CC(=C2)C(=O)O)O
Quantity 1g
Color Brown
Formula Weight 177.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Hydroxyindole-6-carboxylic acid

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold