Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyindole, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
2380-94-1
C8H7NO
133.15
MFCD00005667
NLMQHXUGJIAKTH-UHFFFAOYSA-N
4-hydroxyindole,indol-4-ol,4-indolol,hydroxyindole,4-hydroxy-1h-indole,1h-indole-4-ol,4-hydroxy indole,unii-1w4vd9085v,hydroxyindoles,4-hydroxy-indole
75421
CHEBI:24702
1H-indol-4-ol
C1=CC2=C(C=CN2)C(=C1)O
2380-94-1
C8H7NO
133.15
MFCD00005667
NLMQHXUGJIAKTH-UHFFFAOYSA-N
4-hydroxyindole,indol-4-ol,4-indolol,hydroxyindole,4-hydroxy-1h-indole,1h-indole-4-ol,4-hydroxy indole,unii-1w4vd9085v,hydroxyindoles,4-hydroxy-indole
75421
CHEBI:24702
1H-indol-4-ol
C1=CC2=C(C=CN2)C(=C1)O
Melting Point 97°C to 100°C
Infrared Spectrum Authentic
Color Beige to Gray
Packaging Glass bottle
Quantity 250mg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 21, III, 764
Formula Weight 133.15
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Hydroxyindole, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 219-177-2