Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)phenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
59016-93-2
C7H9BO3
151.956
MFCD00792672
PZRPBPMLSSNFOM-UHFFFAOYSA-N
4-hydroxymethyl phenylboronic acid,4-hydroxymethyl benzeneboronic acid,4-hydroxymethyl phenyl boronic acid,4-hydroxymethylphenyl boronic acid,4-hydroxymethylphenylboronic acid,4-boronobenzyl alcohol,p-hydroxymethyl phenylboronic acid,4-hydroxymethyl phenyl boranediol
2734706
[4-(hydroxymethyl)phenyl]boronic acid
B(C1=CC=C(C=C1)CO)(O)O
59016-93-2
C7H9BO3
151.956
MFCD00792672
PZRPBPMLSSNFOM-UHFFFAOYSA-N
4-hydroxymethyl phenylboronic acid,4-hydroxymethyl benzeneboronic acid,4-hydroxymethyl phenyl boronic acid,4-hydroxymethylphenyl boronic acid,4-hydroxymethylphenylboronic acid,4-boronobenzyl alcohol,p-hydroxymethyl phenylboronic acid,4-hydroxymethyl phenyl boranediol
2734706
[4-(hydroxymethyl)phenyl]boronic acid
B(C1=CC=C(C=C1)CO)(O)O
Color White to Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 151.96
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-(Hydroxymethyl)phenylboronic acid

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Recommended Storage : Refrigerator +4°C