Learn More
4-Hydroxynonenal Mouse anti-Human, ATTO 680, Clone: 12F7, Abnova™
Mouse Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB19287
Catalog No. 89-338-791
Add to cart
Description
Description
4-Hydroxynonenal Monoclonal antibody specifically detects 4-Hydroxynonenal in Human samples. It is validated for WB, ELISA, ICC, IF.
Specifications
Specifications
4-Hydroxynonenal
12F7
Liquid
Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH)
100 μg
Primary
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
ATTO 680
Mouse
IgG1
Store at -20°C
Monoclonal