Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenylacetic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
156-38-7
C8H8O3
152.15
MFCD00004347
XQXPVVBIMDBYFF-UHFFFAOYSA-N
4-hydroxyphenylacetic acid,2-4-hydroxyphenyl acetic acid,p-hydroxyphenylacetic acid,4-hydroxyphenyl acetic acid,4-hydroxyphenylacetate,4-hydroxybenzeneacetic acid,benzeneacetic acid, 4-hydroxy,p-hydroxyphenyl acetic acid,parahydroxy phenylacetic acid,4-carboxymethylphenol
127
CHEBI:18101
2-(4-hydroxyphenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)O
156-38-7
C8H8O3
152.15
MFCD00004347
XQXPVVBIMDBYFF-UHFFFAOYSA-N
4-hydroxyphenylacetic acid,2-4-hydroxyphenyl acetic acid,p-hydroxyphenylacetic acid,4-hydroxyphenyl acetic acid,4-hydroxyphenylacetate,4-hydroxybenzeneacetic acid,benzeneacetic acid, 4-hydroxy,p-hydroxyphenyl acetic acid,parahydroxy phenylacetic acid,4-carboxymethylphenol
127
CHEBI:18101
2-(4-hydroxyphenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)O
Melting Point 147.0°C to 152.0°C
Color Cream to White or Tan
Water 0.5% max. (K.F.)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H4CH2CO2H
Beilstein 10, 190
Solubility Information Solubility in water: soluble in water
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphenylacetic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-851-3

RTECSNumber : AI2680000

TSCA : TSCA