Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
71597-85-8
C6H7BO3
137.93
MFCD01074628
COIQUVGFTILYGA-UHFFFAOYSA-N
4-hydroxyphenyl boronic acid,4-hydroxybenzeneboronic acid,4-boronophenol,4-hydroxyphenyl boranediol,p-hydroxyphenylboronic acid,4-hydroxyphenyl-boronic acid,boronic acid, 4-hydroxyphenyl,pubchem1729,acmc-209ojb
2734360
(4-hydroxyphenyl)boronic acid
B(C1=CC=C(C=C1)O)(O)O
71597-85-8
C6H7BO3
137.93
MFCD01074628
COIQUVGFTILYGA-UHFFFAOYSA-N
4-hydroxyphenyl boronic acid,4-hydroxybenzeneboronic acid,4-boronophenol,4-hydroxyphenyl boranediol,p-hydroxyphenylboronic acid,4-hydroxyphenyl-boronic acid,boronic acid, 4-hydroxyphenyl,pubchem1729,acmc-209ojb
2734360
(4-hydroxyphenyl)boronic acid
B(C1=CC=C(C=C1)O)(O)O
Quantity 5g
Color White to Brown
Melting Point 285.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: 25g/L
Formula Weight 137.93
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Hydroxyphenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C