Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypiperidine, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
5382-16-1
C5H11NO
101.15
MFCD00005999
HDOWRFHMPULYOA-UHFFFAOYSA-N
4-hydroxypiperidine,4-piperidinol,4-hydroxy piperidine,4-hydroxypiperidin,piperidine-4-ol,piperdine-4-ol,piperid-4-ol,4-hydoxypiperidine,4-hydroxypiperdine,4-hyroxypiperidine
79341
piperidin-4-ol
C1CNCCC1O
5382-16-1
C5H11NO
101.15
MFCD00005999
HDOWRFHMPULYOA-UHFFFAOYSA-N
4-hydroxypiperidine,4-piperidinol,4-hydroxy piperidine,4-hydroxypiperidin,piperidine-4-ol,piperdine-4-ol,piperid-4-ol,4-hydoxypiperidine,4-hydroxypiperdine,4-hyroxypiperidine
79341
piperidin-4-ol
C1CNCCC1O
Boiling Point 108.0°C to 114.0°C (10.0 mmHg)
Quantity 500g
Flash Point 107°C
Melting Point 85.0°C to 90.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Beilstein 21,I,188
Solubility Information Solubility in water: soluble.
Formula Weight 101.15
Percent Purity 99+%
Chemical Name or Material 4-Hydroxypiperidine, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-373-1