Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxypropiophenone, 98+%, ACROS Organics™

Click to view available options
Quantity:
500g
Packaging:
Plastic bottle
70-70-2
C9H10O2
150.177
MFCD00002361
RARSHUDCJQSEFJ-UHFFFAOYSA-N
4'-hydroxypropiophenone,paroxypropione,4-hydroxypropiophenone,1-4-hydroxyphenyl propan-1-one,mepal,paroxypropion,frenantol,frenohypon,p-hydroxypropiophenone,proxiphenon
6271
CHEBI:32352
1-(4-hydroxyphenyl)propan-1-one
CCC(=O)C1=CC=C(C=C1)O
70-70-2
C9H10O2
150.177
MFCD00002361
RARSHUDCJQSEFJ-UHFFFAOYSA-N
4'-hydroxypropiophenone,paroxypropione,4-hydroxypropiophenone,1-4-hydroxyphenyl propan-1-one,mepal,paroxypropion,frenantol,frenohypon,p-hydroxypropiophenone,proxiphenon
6271
CHEBI:32352
1-(4-hydroxyphenyl)propan-1-one
CCC(=O)C1=CC=C(C=C1)O
Infrared Spectrum Authentic
Melting Point 147°C to 151°C
Packaging Plastic bottle
Color White
Quantity 500g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula HOC6H4COC2H5
Beilstein 08, 102
Merck Index 15, 7149
Formula Weight 150.18
Physical Form Powder
Percent Purity ≥98.0%
Chemical Name or Material 4';-Hydroxypropiophenone, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-743-2

RTECSNumber : UH1925000

TSCA : TSCA