Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypyridine, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
626-64-2
C5H5NO
95.1
MFCD00006419
GCNTZFIIOFTKIY-UHFFFAOYSA-N
4-hydroxypyridine,pyridin-4-ol,4-pyridinol,4-pyridone,4 1h-pyridinone,pyridin-4 1h-one,4-oxopyridine,4-pyridinone,4-pyridol,gamma-hydroxypyridine
12290
1H-pyridin-4-one
C1=CNC=CC1=O
626-64-2
C5H5NO
95.1
MFCD00006419
GCNTZFIIOFTKIY-UHFFFAOYSA-N
4-hydroxypyridine,pyridin-4-ol,4-pyridinol,4-pyridone,4 1h-pyridinone,pyridin-4 1h-one,4-oxopyridine,4-pyridinone,4-pyridol,gamma-hydroxypyridine
12290
1H-pyridin-4-one
C1=CNC=CC1=O
Density 221g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Flash Point 221°C
Color Beige to Brown
Melting Point 140.0°C to 147.0°C
Boiling Point 230.0°C to 235.0°C (12.0mmHg)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Linear Formula (C5H4N)OH
Beilstein 21, V,7, 152
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol, slightly soluble in chloroform, pract. insoluble in ether and benzene
Formula Weight 95.1
Physical Form Powder or Chunks
Percent Purity 95%
Chemical Name or Material 4-Hydroxypyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 210-958-3