Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
345909-35-5
C10H9NO4
207.185
WZQSVWRLXGIOBK-UHFFFAOYSA-N
4-hydroxyquinoline-2-carboxylic acid hydrate,kynurenic acid hydrate,kynurenic acid monohydrate,4-hydroxyquinaldic acid hydrate,c10h7no3.h2o,2-carboxy-4-hydroxyquinoline hydrate,4-hydroxyquinoline-2-carboxylic acid, hydrate,4-hydroxyquinoline-2-carboxylic acid,hydrate,98,4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
18530299
4-oxo-1H-quinoline-2-carboxylic acid;hydrate
C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O.O
345909-35-5
C10H9NO4
207.185
WZQSVWRLXGIOBK-UHFFFAOYSA-N
4-hydroxyquinoline-2-carboxylic acid hydrate,kynurenic acid hydrate,kynurenic acid monohydrate,4-hydroxyquinaldic acid hydrate,c10h7no3.h2o,2-carboxy-4-hydroxyquinoline hydrate,4-hydroxyquinoline-2-carboxylic acid, hydrate,4-hydroxyquinoline-2-carboxylic acid,hydrate,98,4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
18530299
4-oxo-1H-quinoline-2-carboxylic acid;hydrate
C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O.O
Infrared Spectrum Authentic
Quantity 5g
Melting Point 275.0°C
Solubility Information Solubility in water: 0.9g/L water (100°C). Other solubilities: soluble in hot alcohol, insoluble in ether, very soluble in alkali
Formula Weight 189.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid, hydrate

GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning