Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyquinoline-2-carboxylic Acid, Hydrate 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
345909-35-5
C10H7NO3·xH2O
189.17
MFCD00149476
WZQSVWRLXGIOBK-UHFFFAOYSA-N
4-hydroxyquinoline-2-carboxylic acid hydrate,kynurenic acid hydrate,kynurenic acid monohydrate,4-hydroxyquinaldic acid hydrate,c10h7no3.h2o,2-carboxy-4-hydroxyquinoline hydrate,4-hydroxyquinoline-2-carboxylic acid, hydrate,4-hydroxyquinoline-2-carboxylic acid,hydrate,98,4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
18530299
4-oxo-1H-quinoline-2-carboxylic acid;hydrate
C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O.O
345909-35-5
C10H7NO3·xH2O
189.17
MFCD00149476
WZQSVWRLXGIOBK-UHFFFAOYSA-N
4-hydroxyquinoline-2-carboxylic acid hydrate,kynurenic acid hydrate,kynurenic acid monohydrate,4-hydroxyquinaldic acid hydrate,c10h7no3.h2o,2-carboxy-4-hydroxyquinoline hydrate,4-hydroxyquinoline-2-carboxylic acid, hydrate,4-hydroxyquinoline-2-carboxylic acid,hydrate,98,4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
18530299
4-oxo-1H-quinoline-2-carboxylic acid;hydrate
C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O.O
Color Brown to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 275.0°C
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 22, 230
Merck Index 14, 5327
Solubility Information Solubility in water: 0.9g/L water (100°C). Other solubilities: soluble in hot alcohol, insoluble in ether, very soluble in alkali
Formula Weight 189.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.