Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyquinoline, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
611-36-9
C9H7NO
145.16
MFCD00006777
PMZDQRJGMBOQBF-UHFFFAOYSA-N
4-hydroxyquinoline,quinolin-4-ol,4-quinolinol,quinolin-4 1h-one,4 1h-quinolinone,kynurine,4-hydroxy quinoline,quinoline, 4-hydroxy,ccris 4329,4-chinolinol
69141
CHEBI:15815
1H-quinolin-4-one
C1=CC=C2C(=C1)C(=O)C=CN2
611-36-9
C9H7NO
145.16
MFCD00006777
PMZDQRJGMBOQBF-UHFFFAOYSA-N
4-hydroxyquinoline,quinolin-4-ol,4-quinolinol,quinolin-4 1h-one,4 1h-quinolinone,kynurine,4-hydroxy quinoline,quinoline, 4-hydroxy,ccris 4329,4-chinolinol
69141
CHEBI:15815
1H-quinolin-4-one
C1=CC=C2C(=C1)C(=O)C=CN2
Melting Point 202.0°C to 206.0°C
Color Beige to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 83
Formula Weight 145.16
Physical Form Crystalline Powder or Needles
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning