Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-IBP, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
155798-08-6
C19H21IN2O
420.294
HELCSESNNDZLFM-UHFFFAOYSA-N
4-ibp,n-1-benzylpiperidin-4-yl-4-iodobenzamide,n-n-benzylpiperidin-4-yl-4-iodobenzamide,chembl50112,n-1-benzyl-4-piperidinyl-4-iodobenzamide,bpiba,tocris-0748,125i-n-n-benzylpiperidin-4-yl-4-iodobenzamide,4-iodo-125i-n-1-phenylmethyl-4-piperidinyl benzamide,n-1-benzyl-4-piperidyl-4-iodo-benzamide
132995
N-(1-benzylpiperidin-4-yl)-4-iodobenzamide
C1CN(CCC1NC(=O)C2=CC=C(C=C2)I)CC3=CC=CC=C3
155798-08-6
C19H21IN2O
420.294
HELCSESNNDZLFM-UHFFFAOYSA-N
4-ibp,n-1-benzylpiperidin-4-yl-4-iodobenzamide,n-n-benzylpiperidin-4-yl-4-iodobenzamide,chembl50112,n-1-benzyl-4-piperidinyl-4-iodobenzamide,bpiba,tocris-0748,125i-n-n-benzylpiperidin-4-yl-4-iodobenzamide,4-iodo-125i-n-1-phenylmethyl-4-piperidinyl benzamide,n-1-benzyl-4-piperidyl-4-iodo-benzamide
132995
N-(1-benzylpiperidin-4-yl)-4-iodobenzamide
C1CN(CCC1NC(=O)C2=CC=C(C=C2)I)CC3=CC=CC=C3

σ ligand, σ1 > σ2

Combines high affinity for σ1 and moderate affinity for σ2 sites. Centrally active following systemic administration in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 420.29
Percent Purity >99%
Chemical Name or Material 4-IBP

Recommended Storage : Desiccate at +4°C