Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(Imidazol-1-yl)acetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass Bottle
10041-06-2
C11H10N2O
186.214
MFCD00005282
GAIQQJIMVVUTQN-UHFFFAOYSA-N
4-imidazol-1-yl acetophenone,4'-imidazol-1-yl acetophenone,1-4-1h-imidazol-1-yl phenyl ethanone,4'-1-imidazolyl acetophenone,1-4-imidazol-1-yl-phenyl-ethanone,1-4-acetylphenyl imidazole,ethanone, 1-4-1h-imidazol-1-yl phenyl,1-4-imidazol-1-ylphenyl ethanone,1-4-1h-imidazol-1-yl phenyl ethan-1-one,4'-1h-imidazol-1-yl acetophenone
82316
1-(4-imidazol-1-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2C=CN=C2
10041-06-2
C11H10N2O
186.214
MFCD00005282
GAIQQJIMVVUTQN-UHFFFAOYSA-N
4-imidazol-1-yl acetophenone,4'-imidazol-1-yl acetophenone,1-4-1h-imidazol-1-yl phenyl ethanone,4'-1-imidazolyl acetophenone,1-4-imidazol-1-yl-phenyl-ethanone,1-4-acetylphenyl imidazole,ethanone, 1-4-1h-imidazol-1-yl phenyl,1-4-imidazol-1-ylphenyl ethanone,1-4-1h-imidazol-1-yl phenyl ethan-1-one,4'-1h-imidazol-1-yl acetophenone
82316
1-(4-imidazol-1-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2C=CN=C2
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 112°C to 119°C
Packaging Glass Bottle
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 186.21
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4'-(Imidazol-1-yl)acetophenone, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 233-123-5

TSCA : TSCA