Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Imidazoleacetic acid hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3251-69-2
C5H6N2O2·HCl
162.58
MFCD00012698
MWHLCFYPFGFBQO-UHFFFAOYSA-N
4-imidazoleacetic acid hydrochloride,2-1h-imidazol-5-yl acetic acid hydrochloride,imidazole-4-acetic acid hydrochloride,2-1h-imidazol-4-yl acetic acid hydrochloride,4-imidazoleacetic acid hcl,imidazoleacetic acid hydrochloride,unii-et7a74roao,4-imidazoleaceticacidhydrochloride,i4aa,et7a74roao
145685
2-(1H-imidazol-5-yl)acetic acid;hydrochloride
C1=C(NC=N1)CC(=O)O.Cl
3251-69-2
C5H6N2O2·HCl
162.58
MFCD00012698
MWHLCFYPFGFBQO-UHFFFAOYSA-N
4-imidazoleacetic acid hydrochloride,2-1h-imidazol-5-yl acetic acid hydrochloride,imidazole-4-acetic acid hydrochloride,2-1h-imidazol-4-yl acetic acid hydrochloride,4-imidazoleacetic acid hcl,imidazoleacetic acid hydrochloride,unii-et7a74roao,4-imidazoleaceticacidhydrochloride,i4aa,et7a74roao
145685
2-(1H-imidazol-5-yl)acetic acid;hydrochloride
C1=C(NC=N1)CC(=O)O.Cl
Color White to Yellow
Packaging Glass bottle
Melting Point 221°C to 225°C
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 25, I, 532
Formula Weight 162.58
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Imidazoleacetic acid hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-840-6