Alfa Aesar™ (4-Imidazolyl)acetonitrile, 97%

Manufacturer: Alfa Aesar™ H3299206

Quantity 5g

 View more versions of this product

Catalog No. AAH3299206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18502-05-1
    Molecular Formula C5H5N3
    Formula Weight 107.11
    MDL Number MFCD00070306
    UN Number UN3439
    Synonym 4-(Cyanomethyl)imidazole
    Chemical Name or Material (4-Imidazolyl)acetonitrile

    Specifications

    Formula Weight 107.11
    UN Number UN3439
    Quantity 5g
    Odor Odorless
    Molecular Formula C5H5N3
    MDL Number MFCD00070306
    CAS 18502-05-1
    Synonym 4-(Cyanomethyl)imidazole
    Health Hazard 1 Danger
    Melting Point 136° to 138°C
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material (4-Imidazolyl)acetonitrile
    Assay 97%

    Description

    Applications
    (4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit