Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Indolylboronic acid, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
220465-43-0
C8H8BNO2
160.97
USJUQEVUEBCLLR-UHFFFAOYSA-N
indole-4-boronic acid,4-boronoindole,1h-indol-4-yl boronic acid,1h-indole-4-boronic acid,4-indoleboronic acid,4-indolylboronic acid,4-borono-1h-indole,1h-indol-4-yl-4-boronic acid,boronic acid, 1h-indol-4-yl,b-1h-indol-4-yl-boronic acid
2763173
1H-indol-4-ylboronic acid
B(C1=C2C=CNC2=CC=C1)(O)O
220465-43-0
C8H8BNO2
160.97
USJUQEVUEBCLLR-UHFFFAOYSA-N
indole-4-boronic acid,4-boronoindole,1h-indol-4-yl boronic acid,1h-indole-4-boronic acid,4-indoleboronic acid,4-indolylboronic acid,4-borono-1h-indole,1h-indol-4-yl-4-boronic acid,boronic acid, 1h-indol-4-yl,b-1h-indol-4-yl-boronic acid
2763173
1H-indol-4-ylboronic acid
B(C1=C2C=CNC2=CC=C1)(O)O
Melting Point 184.0°C to 188.0°C
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Formula Weight 160.97
Percent Purity 95%
Chemical Name or Material 4-Indolylboronic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning