Alfa Aesar™ 4-Iodo-2-(trifluoromethyl)benzonitrile, 97%

Manufacturer: Alfa Aesar™ H5560903

Quantity 1g

 View more versions of this product

Catalog No. AAH5560903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 101066-87-9
    Molecular Formula C8H3F3IN
    Formula Weight 297.02
    MDL Number MFCD08458091
    UN Number UN3439
    Synonym 4-Cyano-3-(trifluoromethyl)iodobenzene
    Chemical Name or Material 4-Iodo-2-(trifluoromethyl)benzonitrile

    Specifications

    Formula Weight 297.02
    Molecular Formula C8H3F3IN
    MDL Number MFCD08458091
    UN Number UN3439
    Melting Point 50° to 55°C
    Synonym 4-Cyano-3-(trifluoromethyl)iodobenzene
    Quantity 1g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 101066-87-9
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 4-Iodo-2-(trifluoromethyl)benzonitrile
    Assay 97%

    Description

    Applications
    4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

    Solubility
    Slightly soluble in water.

    Notes
    Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit