Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodo-3,5-dimethylisoxazole, 97%

Click to view available options
Quantity:
1g
5g
10557-85-4
C5H6INO
223.013
MFCD00173743
NMNOXVWRJISEFE-UHFFFAOYSA-N
3,5-dimethyl-4-iodoisoxazole,4-iodo-3,5-dimethylisoxazole,isoxazole, 4-iodo-3,5-dimethyl,4-iodo-3,5-dimethyl-isoxazole,3,5-dimethyl-4-iodoisorazole,pubchem3546,buttpark 97\06-60,acmc-2098gw,ksc496a2p,3.5-dimethyl-4-iodoisorazole
613883
4-iodo-3,5-dimethyl-1,2-oxazole
CC1=C(C(=NO1)C)I
10557-85-4
C5H6INO
223.013
MFCD00173743
NMNOXVWRJISEFE-UHFFFAOYSA-N
3,5-dimethyl-4-iodoisoxazole,4-iodo-3,5-dimethylisoxazole,isoxazole, 4-iodo-3,5-dimethyl,4-iodo-3,5-dimethyl-isoxazole,3,5-dimethyl-4-iodoisorazole,pubchem3546,buttpark 97\06-60,acmc-2098gw,ksc496a2p,3.5-dimethyl-4-iodoisorazole
613883
4-iodo-3,5-dimethyl-1,2-oxazole
CC1=C(C(=NO1)C)I
Sensitivity Light sensitive
Melting Point 51°C to 55°C
Quantity 5g
Formula Weight 223.01
Percent Purity 97%
Chemical Name or Material 4-Iodo-3,5-dimethylisoxazole

TSCA : No

Recommended Storage : Ambient temperatures