Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodo-3-(trifluoromethyl)aniline, 98%

Click to view available options
Quantity:
1g
5g
155403-06-8
C7H5F3IN
287.024
MFCD04972659
ZPVVSYSGZZPGPH-UHFFFAOYSA-N
4-iodo-3-trifluoromethyl aniline,5-amino-2-iodobenzotrifluoride,4-iodo-3-trifluoromethyl-phenylamine,4-iodo-3-trifluoromethyl benzenamine,pubchem2824,3-amino-6-iodo-benzotrifluoride,5-amino-2-iodobenzotrifluoride,5-amino-2-iodobenzotrifluoride,4-iodanyl-3-trifluoromethyl aniline,4-iodo-3-trifluoromethyl aniline, 4-iodo-alpha,alpha,alpha-trifluoro-m-toluidine
23003624
4-iodo-3-(trifluoromethyl)aniline
C1=CC(=C(C=C1N)C(F)(F)F)I
155403-06-8
C7H5F3IN
287.024
MFCD04972659
ZPVVSYSGZZPGPH-UHFFFAOYSA-N
4-iodo-3-trifluoromethyl aniline,5-amino-2-iodobenzotrifluoride,4-iodo-3-trifluoromethyl-phenylamine,4-iodo-3-trifluoromethyl benzenamine,pubchem2824,3-amino-6-iodo-benzotrifluoride,5-amino-2-iodobenzotrifluoride,5-amino-2-iodobenzotrifluoride,4-iodanyl-3-trifluoromethyl aniline,4-iodo-3-trifluoromethyl aniline, 4-iodo-alpha,alpha,alpha-trifluoro-m-toluidine
23003624
4-iodo-3-(trifluoromethyl)aniline
C1=CC(=C(C=C1N)C(F)(F)F)I
Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and it is also an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light. Store in dark.
Quantity 5g
Sensitivity Light Sensitive
Color White
UN Number UN2811
Beilstein 11440140
Formula Weight 287.02
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)aniline

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.