Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodo-D-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
25g
62561-75-5
C9H10INO2
291.088
MFCD00063067
PZNQZSRPDOEBMS-MRVPVSSYSA-N
4-iodo-d-phenylalanine,h-d-phe 4-i-oh,d-4-iodophenylalanine,d-phe 4-i-oh,p-iodo-d-phenylalanine,d-4-iodophe,r-2-amino-3-4-iodophenyl propanoic acid,d-4-iodo phenylalanine,2r-2-amino-3-4-iodophenyl propanoic acid,d-4-iodophenyl alanine
2733279
(2R)-2-amino-3-(4-iodophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)I
62561-75-5
C9H10INO2
291.088
MFCD00063067
PZNQZSRPDOEBMS-MRVPVSSYSA-N
4-iodo-d-phenylalanine,h-d-phe 4-i-oh,d-4-iodophenylalanine,d-phe 4-i-oh,p-iodo-d-phenylalanine,d-4-iodophe,r-2-amino-3-4-iodophenyl propanoic acid,d-4-iodo phenylalanine,2r-2-amino-3-4-iodophenyl propanoic acid,d-4-iodophenyl alanine
2733279
(2R)-2-amino-3-(4-iodophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)I
Formula Weight 291.09
Physical Form Powder
Percent Purity 95%
Color Yellow
Melting Point 251°C
Quantity 1g
Sensitivity Light Sensitive
Chemical Name or Material 4-Iodo-D-phenylalanine

storageNote1 : 2° to 8°C