Alfa Aesar™ 4-Iodo-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H51979MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51979MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24250-85-9
    Molecular Formula C9H10INO2
    Formula Weight 291.09
    MDL Number MFCD00002602
    Beilstein 4411317
    Synonym (S)-2-Amino-3-(4-iodophenyl)propionic acid; H-Phe(4-I)-OH
    Chemical Name or Material 4-Iodo-L-phenylalanine

    Specifications

    Formula Weight 291.09
    Molecular Formula C9H10INO2
    MDL Number MFCD00002602
    CAS 24250-85-9
    Beilstein 4411317
    Synonym (S)-2-Amino-3-(4-iodophenyl)propionic acid; H-Phe(4-I)-OH
    Quantity 250mg
    Optical Rotation −6.5° (c=1 in Acetic Acid:Water (4:1))
    Chemical Name or Material 4-Iodo-L-phenylalanine
    Assay 95%

    Description

    Applications
    4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

    Solubility
    Partly miscible with water.

    Notes
    Light sensitive. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit