Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodo-o-phenylenediamine, 95%

Click to view available options
Quantity:
250mg
1g
5g
21304-38-1
C6H7IN2
234.04
MFCD08669466
FUOSRKZBOIVBOS-UHFFFAOYSA-N
1,2-diamino-4-iodobenzene,4-iodo-1,2-diaminobenzene,4-iodo-1,2-phenylenediamine,1,2-benzenediamine, 4-iodo,4-iodo-1,2-benzenediamine,4-iodo-benzene-1,2-diamine,4-iodo-1,2-phenylenediamine,ambkkkkk335,2-amino-5-iodoaniline,2-amino4-iodophenylamine
11207049
4-iodobenzene-1,2-diamine
C1=CC(=C(C=C1I)N)N
21304-38-1
C6H7IN2
234.04
MFCD08669466
FUOSRKZBOIVBOS-UHFFFAOYSA-N
1,2-diamino-4-iodobenzene,4-iodo-1,2-diaminobenzene,4-iodo-1,2-phenylenediamine,1,2-benzenediamine, 4-iodo,4-iodo-1,2-benzenediamine,4-iodo-benzene-1,2-diamine,4-iodo-1,2-phenylenediamine,ambkkkkk335,2-amino-5-iodoaniline,2-amino4-iodophenylamine
11207049
4-iodobenzene-1,2-diamine
C1=CC(=C(C=C1I)N)N
Applications
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 73°C to 77°C
Sensitivity Light Sensitive
Quantity 5g
UN Number UN2811
Beilstein 2716284
Formula Weight 234.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Iodo-o-phenylenediamine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.