Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Iodo-o-phenylenediamine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 73°C to 77°C |
Quantity | 250 mg |
UN Number | UN2811 |
Beilstein | 2716284 |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 234.04 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 4-Iodo-o-phenylenediamine |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction