Alfa Aesar™ 4-Iodoaniline, 98%

Manufacturer: Alfa Aesar™ B2193630

Quantity 250g

 View more versions of this product

Catalog No. AAB2193630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 540-37-4
    Molecular Formula C6H6IN
    Formula Weight 219.03
    MDL Number MFCD00007848
    Beilstein 774101
    Merck Index 14,5027
    Chemical Name or Material 4-Iodoaniline

    Specifications

    Formula Weight 219.03
    TSCA TSCA
    MDL Number MFCD00007848
    Health Hazard 1 Danger
    Melting Point 61° to 64°C
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C6H6IN
    CAS 540-37-4
    Beilstein 774101
    Merck Index 14,5027
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number BY3850000
    Chemical Name or Material 4-Iodoaniline
    Assay 98%

    Description

    Applications
    4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

    Solubility
    Slightly soluble in water.Soluble in chloroform, methanol.

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

    GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit