Alfa Aesar™ 4-Iodoanisole, 98+%

Manufacturer: Alfa Aesar™ A1214436

Quantity 500g

 View more versions of this product

Catalog No. AAA1214436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 696-62-8
    Molecular Formula C7H7IO
    Formula Weight 234.04
    MDL Number MFCD00001056
    Beilstein 1906692
    Synonym 4-Methoxyiodobenzene
    Chemical Name or Material 4-Iodoanisole

    Specifications

    Formula Weight 234.04
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00001056
    CAS 696-62-8
    Synonym 4-Methoxyiodobenzene
    Beilstein 1906692
    Health Hazard 1 Warning
    Melting Point 48° to 52°C
    Quantity 500g
    Molecular Formula C7H7IO
    Boiling Point 240°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point >110°C (230°F)
    Chemical Name or Material 4-Iodoanisole
    Assay ≥98%

    Description

    Applications
    4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

    Solubility
    Soluble in ethanol, ether, chloroform.

    Notes
    Light sensitive. Store in the dark. Store away from oxidizing agents, light.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit