Alfa Aesar™ 4-Iodobenzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H2819006

Quantity 5g

 View more versions of this product

Catalog No. AAH2819006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73852-88-7
    Molecular Formula C12H16BIO2
    Formula Weight 329.97
    MDL Number MFCD09953499
    Synonym 4-Iodophenylboronic acid pinacol ester; 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 4-Iodobenzeneboronic acid pinacol ester

    Specifications

    Formula Weight 329.97
    Molecular Formula C12H16BIO2
    Quantity 5g
    MDL Number MFCD09953499
    CAS 73852-88-7
    Melting Point 102° to 104°C
    Synonym 4-Iodophenylboronic acid pinacol ester; 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 4-Iodobenzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit