Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodobenzenesulfonamide, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
825-86-5
C6H6INO2S
283.09
KQZFABSTXSNEQH-UHFFFAOYSA-N
p-iodobenzenesulfonamide,4-iodobenzenesulphonamide,4-iodobenzene-1-sulfonamide,benzenesulfonamide,4-iodo,benzenesulfonamide, p-iodo,p-jodbenzolsulfonamid,4-iodibenzenesulphonamide,4-iodo-benzenesulfonamide,4-iodo-1-sulfamoylbenzene,3-11-00-00110 beilstein handbook reference
13218
4-iodobenzenesulfonamide
C1=CC(=CC=C1S(=O)(=O)N)I
825-86-5
C6H6INO2S
283.09
KQZFABSTXSNEQH-UHFFFAOYSA-N
p-iodobenzenesulfonamide,4-iodobenzenesulphonamide,4-iodobenzene-1-sulfonamide,benzenesulfonamide,4-iodo,benzenesulfonamide, p-iodo,p-jodbenzolsulfonamid,4-iodibenzenesulphonamide,4-iodo-benzenesulfonamide,4-iodo-1-sulfamoylbenzene,3-11-00-00110 beilstein handbook reference
13218
4-iodobenzenesulfonamide
C1=CC(=CC=C1S(=O)(=O)N)I
Packaging Glass Bottle
Infrared Spectrum Authentic
Loss on Drying 1% max.
Melting Point 193.0°C to 195.0°C
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Linear Formula IC6H4SO2NH2
Formula Weight 283.09
Percent Purity 95%
Chemical Name or Material 4-Iodobenzenesulfonamide

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning