Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Iodobenzenesulfonyl chloride, 97%

Click to view available options
Quantity:
5g
25g
100g
98-61-3
C6H4ClIO2S
302.51
MFCD00007441
POXFXTSTVWDWIR-UHFFFAOYSA-N
pipsyl chloride,4-iodobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-iodo,p-iodobenzenesulfonyl chloride,4-iodobenzenesulphonyl chloride,4-iodobenzenesulfonylchloride,4-iodophenylsulfonyl chloride,iodobenzene-p-sulfonyl chloride,unii-mvl274ezog,mvl274ezog
7399
4-iodobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)I
98-61-3
C6H4ClIO2S
302.51
MFCD00007441
POXFXTSTVWDWIR-UHFFFAOYSA-N
pipsyl chloride,4-iodobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-iodo,p-iodobenzenesulfonyl chloride,4-iodobenzenesulphonyl chloride,4-iodobenzenesulfonylchloride,4-iodophenylsulfonyl chloride,iodobenzene-p-sulfonyl chloride,unii-mvl274ezog,mvl274ezog
7399
4-iodobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)I
Applications
It is used as intermediate in organic syntheses & in medicine.

Solubility
Reacts with water.

Notes
Moisture & light sensitive. Store away from oxidizing agent.
Sensitivity Light and moisture sensitive
Quantity 5g
Melting Point 79°C to 84°C
UN Number UN3261
Beilstein 2691661
Merck Index 14,7486
Solubility Information Reacts with water.
Formula Weight 302.52
Percent Purity 97%
Chemical Name or Material 4-Iodobenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 202-686-9

TSCA : Yes

Recommended Storage : Ambient temperatures