Alfa Aesar™ 4-Iodobenzonitrile, 98%

Manufacturer: Alfa Aesar™ B2510406

Quantity 5g

 View more versions of this product

Catalog No. AAB2510406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3058-39-7
    Molecular Formula C7H4IN
    Formula Weight 229.02
    MDL Number MFCD00051310
    UN Number UN3439
    Synonym 4-Cyanoiodobenzene
    Chemical Name or Material 4-Iodobenzonitrile

    Specifications

    Formula Weight 229.02
    Molecular Formula C7H4IN
    UN Number UN3439
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00051310
    CAS 3058-39-7
    Melting Point 125° to 127°C
    Synonym 4-Cyanoiodobenzene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 4-Iodobenzonitrile
    Assay 98%

    Description

    Applications
    4-Iodobenzonitrile is used in organic synthesis. It is used as catalytic agent, petrochemical additive.

    Solubility
    Insoluble in water.

    Notes
    Light Sensitive. Store in the dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

    GHS H Statement
    H301-H312-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit