Alfa Aesar™ 4-Iodobenzotrifluoride, 98%

Manufacturer: Alfa Aesar™ A1218906

Quantity 5g

 View more versions of this product

Catalog No. AAA1218906

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 455-13-0
    Molecular Formula C7H4F3I
    Formula Weight 272.01
    MDL Number MFCD00039398
    Beilstein 1944013
    Synonym 4-Iodo-alpha,alpha,alpha-trifluorotoluene
    Chemical Name or Material 4-Iodobenzotrifluoride

    Specifications

    Formula Weight 272.01
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H4F3I
    CAS 455-13-0
    Flash Point 68°C (154°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    TSCA TSCA
    MDL Number MFCD00039398
    Boiling Point 68° to 70°C (15mmHg)
    Synonym 4-Iodo-alpha,alpha,alpha-trifluorotoluene
    Beilstein 1944013
    Melting Point −8°C
    Quantity 5g
    Chemical Name or Material 4-Iodobenzotrifluoride
    Assay 98%

    Description

    Applications
    4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

    Solubility
    Insoluble in water.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents. Store in dark.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit